Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Calreticulin Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC38981V100UG
This item is not returnable.
View return policy
Description
Calreticulin Monoclonal specifically detects Calreticulin in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Calreticulin | |
Monoclonal | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
autoantigen Ro, CALR, Calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA, Vasostatin | |
E. coli-derived recombinant human Calreticulin, Glu18-Leu417, Accession # P27797 | |
100 μg | |
Primary | |
Detects human Calreticulin in ELISA. Detects human, mouse and rat Calreticulin in Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2b |
Flow Cytometry | |
681233 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P27797 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
811 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction