Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Calreticulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
$188.28 - $472.87
Specifications
| Antigen | Calreticulin |
|---|---|
| Clone | 681207 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB38982SP
![]() |
R&D Systems
MAB38982SP |
25 μg |
Each for $188.28
|
|
|||||
MAB38982100
![]() |
R&D Systems
MAB38982100 |
100 μg |
Each for $472.87
|
|
|||||
Description
Calreticulin Monoclonal specifically detects Calreticulin in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Calreticulin | |
| Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL | |
| Monoclonal | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| autoantigen Ro, CALR, Calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA, Vasostatin | |
| E. coli-derived recombinant human Calreticulin, Glu18-Leu417, Accession # P27797 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Calreticulin in direct ELISAs. In direct ELISAs, no cross-reactivity with human Vasostatin is observed. |
| 681207 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Human | |
| P27797 | |
| 811 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title