Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Calsyntenin-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5480
This item is not returnable.
View return policy
Description
Calsyntenin-2 Polyclonal specifically detects Calsyntenin-2 in Human samples. It is validated for Immunohistochemistry.Specifications
Calsyntenin-2 | |
Polyclonal | |
Unconjugated | |
Q9H4D0 | |
CLSTN2 | |
Mouse myeloma cell line NS0-derived recombinant human Calsyntenin-2 Ser21-Thr834 (Val366Ile) Accession # Q9H4D0 | |
100 μg | |
Primary | |
Detects human Calsyntenin-2 in direct ELISAs. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Calsyntenin-1 and rhCalsyntenin-3 is observed. | |
Human | |
IgG |
Immunohistochemistry | |
LYOPH | |
Immunohistochemistry 5-15 ug/mL | |
alcadein gamma, Alcadein-gamma, alcagamma, ALC-GAMMA, cadherin-related family member 13, calsyntenin 2, calsyntenin-2, Calsyntenin2, CDHR13, CLSTN2, CS2, CS2alcadein-gamma, CSTN2, FLJ39113, FLJ39499, MGC119560 | |
Sheep | |
Affinity Purified | |
RUO | |
64084 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction