Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CaM Kinase II delta Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB4176SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4176SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB4176SP Supplier R&D Systems Supplier No. MAB4176SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CaM Kinase II delta Monoclonal specifically detects CaM Kinase II delta in Human samples. It is validated for Western Blot.

Specifications

Antigen CaM Kinase II delta
Applications Western Blot
Classification Monoclonal
Clone 438422
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13557
Gene Alias calcium/calmodulin-dependent protein kinase (CaM kinase) II delta, calcium/calmodulin-dependent protein kinase II delta, calcium/calmodulin-dependent protein kinase type II delta chain, calcium/calmodulin-dependent protein kinase type II subunit delta, CaM kinase II delta subunit, CaM kinase II subunit delta, CAMK2D, CAMKD, CaMK-II delta subunit, CaMK-II subunit delta, CaM-kinase II delta chain, DKFZp686G23119, DKFZp686I2288, EC 2.7.11, EC 2.7.11.17, MGC44911
Gene Symbols CAMK2D
Host Species Mouse
Immunogen E. coli-derived recombinant human CaM Kinase II delta Ala402-Lys478 Accession # Q13557
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 817
Test Specificity Detects endogenous human CaM Kinase II delta isoforms in Western blots. In Western blots, this antibody does not cross-react with recombinant human CaMKII alpha , CaMKII beta , or CaMKII gamma .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.