Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CAMP/LL37/FALL39 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7497
This item is not returnable.
View return policy
Description
CAMP/LL37/FALL39/Cathelicidin Polyclonal specifically detects CAMP/LL37/FALL39/Cathelicidin in Human samples. It is validated for Western Blot.Specifications
CAMP/LL37/FALL39/Cathelicidin | |
Polyclonal | |
Unconjugated | |
P49913 | |
CAMP | |
E. coli-derived recombinant human CAMP/LL37/FALL39 Leu134-Ser170 Accession # P49913 | |
100 μg | |
Primary | |
Detects human CAMP/LL37/FALL39 in direct ELISAs and Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
18 kDa cationic antimicrobial protein, CAMP, CAP18, CAP18CRAMP, cathelicidin antimicrobial peptide, CRAMP, FALL39, FALL-39, FALL39CAP-18, hCAP-18, LL37 | |
Sheep | |
Affinity Purified | |
RUO | |
820 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction