Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Canine IL-23 p19 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6250
This item is not returnable.
View return policy
Description
IL-23A/IL-23 P19 Polyclonal specifically detects IL-23A/IL-23 P19 in Human, Canine samples. It is validated for Western Blot.Specifications
| IL-23A/IL-23 P19 | |
| Polyclonal | |
| Unconjugated | |
| XP_538231 | |
| IL23A | |
| E. coli-derived recombinant canine IL-23 p19 Arg23-Pro193 Accession # XP_538231 | |
| 100 μg | |
| Primary | |
| Detects human and canine IL-23 p19 in direct ELISAs and Western blots. In direct ELISAs, approximately 60% cross-reactivity with recombinant feline IL-23 p19 is observed and approximately 40% cross-reactivity with recombinant mouse IL-23 p19 is observed. | |
| Human, Canine | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| IL23, IL-23, IL-23 p19, IL-23 p19/IL-12 p40, IL23A, IL-23A, IL-23-A, IL-23p19, IL-23p19/IL-12p40, IL23P19P19, interleukin 23 p19 subunit, interleukin 23, alpha subunit p19, interleukin-23 subunit alpha, Interleukin-23 subunit p19, JKA3 induced upon T-cell activation, MGC79388, SGRF, SGRFIL-23 subunit alpha | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 51561 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction