Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Canine NGFR/TNFRSF16 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB367
This item is not returnable.
View return policy
Description
NGFR/TNFRSF16/p75NTR Monoclonal specifically detects NGFR/TNFRSF16/p75NTR in Human, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
NGFR/TNFRSF16/p75NTR | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P08138 | |
NGFR | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human NGF R Lys29-Asn250 Accession # P08138 | |
100 μg | |
Primary | |
Detects human NGF R in direct ELISAs and Western blots. In direct ELISAs, no crossreactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD40, rhBAFF R, rhCD30, rhDR3, rhDR6, rhEDAR, rhFas, rhHVEM, rhGITR, rhLTR B, recominant mouse (rm) NGF R, rhOPG, rmOX40, rhRANK, rhTAJ, rhTNF RI or rhTNF RII is observed. | |
Human, Canine | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
74902 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD271, CD271 antigen, Gp80-LNGFR, member 16, nerve growth factor receptor, NGF R, NGF receptor, p75 ICD, p75 NTR, TNFRSF16, TNFRSF16nerve growth factor receptor (TNFR superfamily, member 16), tumor necrosis factor receptor superfamily member 16 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4804 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction