Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cannabinoid R1/CB1/CNR1 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB3834T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3834T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3834T100 Supplier R&D Systems Supplier No. FAB3834T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cannabinoid R1/CB1/CNR1 Monoclonal specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Cannabinoid R1/CB1/CNR1
Applications Flow Cytometry
Classification Monoclonal
Clone 368302
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. NP_057167
Gene Alias CANN6, CANN6CB1R, cannabinoid receptor 1 (brain), CannabinoidR1, CB1, CB1A, CB1K5, CB1R, CB-R, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR, CNR1, SKR6R
Gene Symbols CNR1
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1 Met1-Leu472 Accession # NP_057167
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1268
Test Specificity Stains Cannabinoid R1/CB1/CNR1-transfected cells but not irrelevant transfectants.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.