Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cannabinoid R1/CB1/CNR1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3834
This item is not returnable.
View return policy
Description
Cannabinoid R1/CB1/CNR1 Monoclonal specifically detects Cannabinoid R1/CB1/CNR1 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Cannabinoid R1/CB1/CNR1 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
NP_057167 | |
CNR1 | |
HEK293 human embryonic kidney cell line transfected with human Cannabinoid R1/CB1/CNR1 Met1-Leu472 Accession # NP_057167 | |
100 μg | |
Primary | |
Stains Cannabinoid R1/CB1/CNR1-transfected cells but not irrelevant transfectants. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
368302 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CANN6, CANN6CB1R, cannabinoid receptor 1 (brain), CannabinoidR1, CB1, CB1A, CB1K5, CB1R, CB-R, CB-RCB1cannabinoid receptor 1, central cannabinoid receptor, CNR, CNR1, SKR6R | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1268 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction