Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cannabinoid R2/CB2/CNR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB36551R100
This item is not returnable.
View return policy
Description
Cannabinoid R2/CB2/CNR2 Monoclonal specifically detects Cannabinoid R2/CB2/CNR2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
Cannabinoid R2/CB2/CNR2 | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
cannabinoid receptor 2 (macrophage), CannabinoidR2, CB-2, CB2cannabinoid receptor 2, CB2R, CNR2, CX5, CX5testis-dominant CNR2 isoform CB2, hCB2 | |
Human embryonic kidney cell line HEK293-derived transfected with human Cannabinoid R2/CB2/CNR2, Met1-Cys360, Accession # NP_001832 | |
100 μg | |
1269 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
IgG2b |
Flow Cytometry, CyTOF | |
352110R | |
Flow Cytometry 0.25 μg/106 cells, CyTOF-ready | |
NP_001832 | |
Mouse | |
Protein A or G purified from cell culture supernatant | |
Primary | |
Detects human Cannabinoid R2/CB2/CNR2 in HEK293 human cell line transfected with human Cannabinoid R2/CB2/CNR2 and not Cannabinoid R1/CB1/CNR1 transfectants in flow cytometry. | |
Human | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction