Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbohydrate Sulfotransferase 1/CHST1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5316
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 1/CHST1/KS6ST Polyclonal specifically detects Carbohydrate Sulfotransferase 1/CHST1/KS6ST in Human samples. It is validated for Western Blot.Specifications
Carbohydrate Sulfotransferase 1/CHST1/KS6ST | |
Polyclonal | |
Unconjugated | |
O43916 | |
CHST1 | |
Chinese hamster ovary cell line CHO-derived recombinant human Carbohydrate Sulfotransferase 1/CHST1 Arg24-Ser411 Accession # O43916 | |
100 μg | |
Primary | |
Detects human Carbohydrate Sulfotransferase 1/CHST1 in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse (rm) CHST1 is observed and less than 5% cross-reactivity with recombinant human (rh) CHST2, rhCHST5, and rmCHST7 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
6-O-sulfotransferase 1, C6ST, carbohydrate (chondroitin 6/keratan) sulfotransferase 1, carbohydrate (keratan sulfate Gal-6) sulfotransferase 1, carbohydrate sulfotransferase 1, CHST1, EC 2.8.2, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 1, GST-1, Keratan sulfate Gal-6 sulfotransferase, KS6ST, KSGal6ST, KSGal6STEC 2.8.2.21, KSST | |
Sheep | |
Affinity Purified | |
RUO | |
8534 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction