Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbohydrate Sulfotransferase 1/CHST1 Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF5316
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF5316 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF5316 Supplier R&D Systems Supplier No. BAF5316
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Carbohydrate Sulfotransferase 1/CHST1/KS6ST Polyclonal specifically detects Carbohydrate Sulfotransferase 1/CHST1/KS6ST in Human samples. It is validated for Western Blot.

Specifications

Antigen Carbohydrate Sulfotransferase 1/CHST1/KS6ST
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. O43916
Gene Alias 6-O-sulfotransferase 1, C6ST, carbohydrate (chondroitin 6/keratan) sulfotransferase 1, carbohydrate (keratan sulfate Gal-6) sulfotransferase 1, carbohydrate sulfotransferase 1, CHST1, EC 2.8.2, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 1, GST-1, Keratan sulfate Gal-6 sulfotransferase, KS6ST, KSGal6ST, KSGal6STEC 2.8.2.21, KSST
Gene Symbols CHST1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Carbohydrate Sulfotransferase 1/CHST1 Arg24-Ser411 Accession # O43916
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8534
Test Specificity Detects human Carbohydrate Sulfotransferase 1/CHST1 in Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse CHST1 is observed and less than 5% cross-reactivity with recombinant human (rh) CHST2, rhCHST5, and rmCHST7 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.