Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbohydrate Sulfotransferase 2/CHST2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5107
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 2/CHST2 Polyclonal specifically detects Carbohydrate Sulfotransferase 2/CHST2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carbohydrate Sulfotransferase 2/CHST2 | |
Polyclonal | |
Unconjugated | |
Q9Y4C5 | |
CHST2 | |
Chinese hamster ovary cell line CHO-derived recombinant human CHST2 Tyr76-Leu530 Accession # Q9Y4C5 | |
100 μg | |
Primary | |
Detects human Carbohydrate Sulfotransferase 2/CHST2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) CHST1, rhCHST5, and recombinant mouse CHST7 is observed. | |
Human | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
C6ST, carbohydrate (chondroitin 6/keratan) sulfotransferase 2, carbohydrate (N-acetylglucosamine-6-O) sulfotransferase 2, carbohydrate sulfotransferase 2, CHST2, EC 2.8.2.-, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 2, glcNAc6ST-1, GN6ST, Gn6ST-1, GST2, GST-2, N-acetylglucosamine 6-O-sulfotransferase 1 | |
Goat | |
Affinity Purified | |
RUO | |
9435 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction