Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbohydrate Sulfotransferase 3/CHST3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB7450SP
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 3/CHST3 Monoclonal specifically detects Carbohydrate Sulfotransferase 3/CHST3 in Human samples. It is validated for Immunohistochemistry.Specifications
Carbohydrate Sulfotransferase 3/CHST3 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 8-25 ug/mL | |
Q7LGC8 | |
CHST3 | |
Chinese hamster ovary cell line CHO-derived recombinant human Carbohydrate Sulfotransferase 3/CHST3 Glu39-Thr479 Accession # Q7LGC8 | |
25 μg | |
Primary | |
Detects human Carbohydrate Sulfotransferase 3/CHST3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CHST1 or recombinant mouse CHST3 is observed. | |
Human | |
Purified |
Immunohistochemistry | |
799011 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C6ST1, C6ST-1, C6ST1HSD, C6STEC 2.8.2.17, carbohydrate (chondroitin 6) sulfotransferase 3, carbohydrate sulfotransferase 3, Chondroitin 6-O-sulfotransferase 1, Chondroitin 6-sulfotransferase, CHST3, EC 2.8.2, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0, GST-0 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9469 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction