Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbohydrate Sulfotransferase 4/CHST4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5357SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5357SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5357SP Supplier R&D Systems Supplier No. AF5357SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Carbohydrate Sulfotransferase 4/CHST4 Polyclonal specifically detects Carbohydrate Sulfotransferase 4/CHST4 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.

Specifications

Antigen Carbohydrate Sulfotransferase 4/CHST4
Applications Western Blot, Immunoprecipitation, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 10.7 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NCG5
Gene Alias carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4, CHST4, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3, GlcNAc6ST2, GlcNAc6ST-2, Gn6st-2, GST3, GST-3, HEC-GlcNAc6ST, HECGLCNAC6ST, HEC-GLCNAC-6-ST, High endothelial cells N-acetylglucosamine 6-O-sulfotransferase, L-selectin ligand sulfotransferase, LSST, LSSTcarbohydrate sulfotransferase 4, N-acetylglucosamine 6-O-sulfotransferase 2
Gene Symbols CHST4
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Sulfotransferase 4/CHST4 Leu22-Gln286 Accession # Q8NCG5
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10164
Test Specificity Detects human Sulfotransferase 4/CHST4 in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant mouse CHST4 is observed and less than 5% cross-reactivity with recombinant human (rh) CHST1, rhCHST5, and rhCHST6 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.