Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbohydrate Sulfotransferase 4/CHST4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5357
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5357 100 μg
MAB5357SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB5357 Supplier R&D Systems Supplier No. MAB5357
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carbohydrate Sulfotransferase 4/CHST4 Monoclonal specifically detects Carbohydrate Sulfotransferase 4/CHST4 in Human samples. It is validated for Immunoprecipitation, Neutralization.

Specifications

Antigen Carbohydrate Sulfotransferase 4/CHST4
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 595223
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation, Neutralization
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q8NCG5
Gene Alias carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4, CHST4, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3, GlcNAc6ST2, GlcNAc6ST-2, Gn6st-2, GST3, GST-3, HEC-GlcNAc6ST, HECGLCNAC6ST, HEC-GLCNAC-6-ST, High endothelial cells N-acetylglucosamine 6-O-sulfotransferase, L-selectin ligand sulfotransferase, LSST, LSSTcarbohydrate sulfotransferase 4, N-acetylglucosamine 6-O-sulfotransferase 2
Gene Symbols CHST4
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Sulfotransferase 4/CHST4 His29-His386 Accession # Q8NCG5
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10164
Test Specificity Detects human Carbohydrate Sulfotransferase 4/CHST4 is direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human CHST1, 2, 5, 6, 10, 14, recombinant mouse CHST3, 4, and 7 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.