Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbonic Anhydrase I/CA1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2180
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2180 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB2180 Supplier R&D Systems Supplier No. MAB2180
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Carbonic Anhydrase I/CA1 Monoclonal specifically detects Carbonic Anhydrase I/CA1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Carbonic Anhydrase I/CA1
Applications Western Blot
Classification Monoclonal
Clone 363121
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P00915
Gene Alias CA1, CA-I, Car1, Carbonate dehydratase I, carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase ICAB, carbonic dehydratase, EC 4.2.1.1
Gene Symbols CA1
Host Species Rat
Immunogen E. coli-derived recombinant human Carbonic Anhydrase I/CA1 Ala2-Phe261 Accession # P00915
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 759
Test Specificity Detects human Carbonic Anhydrase I/CA1 in direct ELISAs and Western blots. In Western blots, 30% cross-reactivity with recombinant human (rh) CA2 is observed and no cross-reactivity with rhCA3, -4, -7, -8, -10, -11, -12, -13, or -14 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.