Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbonic Anhydrase III/CA3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2185
This item is not returnable.
View return policy
Description
Carbonic Anhydrase III/CA3 Monoclonal specifically detects Carbonic Anhydrase III/CA3 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carbonic Anhydrase III/CA3 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
P07451 | |
CA3 | |
E. coli-derived recombinant human Carbonic Anhydrase III/CA3 Ala2-Lys260 Accession # P07451 | |
100 μg | |
Primary | |
Detects human Carbonic Anhydrase III/CA3 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human CA1, 2, 4, 5a, 5b, 6, 7, 8, or 9 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
495018 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CA3, CAIII, CA-III, Car3, Carbonate dehydratase III, carbonic anhydrase 3, Carbonic anhydrase III, carbonic anhydrase III, muscle specific, EC 4.2.1.1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
761 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction