Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbonic Anhydrase IV/CA4 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB2186
This item is not returnable.
View return policy
Description
Carbonic Anhydrase IV/CA4 Monoclonal specifically detects Carbonic Anhydrase IV/CA4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Carbonic Anhydrase IV/CA4 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
P22748 | |
CA4 | |
Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase IV/CA4 Ala19-Lys283 Accession # P22748 | |
100 μg | |
Primary | |
Detects human Carbonic Anhydrase IV/CA4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 10% cross-reactivity with recombinant human (rh) CA1, rhCA2, and recombinant mouse CA4 is observed. Does not cross-react with rhCA3, 8, 9, 10, 12, 13, or 14. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
310413 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CA4, CAIV, CA-IV, Car4, Carbonate dehydratase IV, carbonic anhydrase 4, carbonic anhydrase IVRP17, carbonic dehydratase IV, EC 4.2.1.1, retinitis pigmentosa 17 (autosomal dominant), RP17 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
762 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction