Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbonic Anhydrase VB/CA5B Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3176
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VB/CA5B Polyclonal specifically detects Carbonic Anhydrase VB/CA5B in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Carbonic Anhydrase VB/CA5B | |
Polyclonal | |
Unconjugated | |
Q9Y2D0 | |
CA5B | |
E. coli-derived recombinant human Carbonic Anhydrase VB/CA5B Cys34-Pro317 Accession # Q9Y2D0 | |
100 μg | |
Primary | |
Detects human Carbonic Anhydrase VB/CA5B in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) CA-5A, rhCA-2, and rhCA-7 is observed. | |
Human | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
CA5B, Carbonate dehydratase VB, carbonic anhydrase 5B, mitochondrial, Carbonic anhydrase VB, carbonic anhydrase VB, mitochondrial, carbonic dehydratase, CAVB, CA-VB, EC 4.2.1.1, MGC39962 | |
Goat | |
Affinity Purified | |
RUO | |
11238 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction