Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbonic Anhydrase VI Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2939
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2939 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2939 Supplier R&D Systems Supplier No. AF2939
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carbonic Anhydrase VI Polyclonal specifically detects Carbonic Anhydrase VI in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.

Specifications

Antigen Carbonic Anhydrase VI
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 1 ug/mL, Immunoprecipitation 25 ug/mL
Gene Accession No. EAW71606
Gene Alias CA6, Carbonate dehydratase VI, carbonic anhydrase 6, carbonic anhydrase VIGUSTIN, CA-VI, EC 4.2.1.1, Gustin, MGC21256, Salivary carbonic anhydrase, Secreted carbonic anhydrase
Gene Symbols CA6
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase VI Gln18-Asn308 Accession # EAW71606
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 765
Test Specificity Detects human Carbonic Anhydrase VI in direct ELISAs and Western blots. In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) Carbonic Anhydrase (CA) 5A and rhCA5B is observed and less than 1% cross-reactivity with rhCA1, rhCA2, rhCA3, rhCA4, rhCA7, rhCA8, rhCA9, rhCA10, rhCA12, rhCA13, and rhCA14 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.