Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carbonic Anhydrase XII/CA12 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2190SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XII/CA12 Polyclonal specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Western Blot, Simple Western.Specifications
Carbonic Anhydrase XII/CA12 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CA12, Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3 | |
Goat | |
Affinity Purified | |
RUO | |
771 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
O43570 | |
CA12 | |
Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase XII/CA12 Ala25-Gln291 Accession # O43570 | |
25 μg | |
Primary | |
Detects human Carbonic Anhydrase XII/CA12 in direct ELISAs and Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse Carbonic Anhydrase XII/CA12 is observed and less than 5% cross-reactivity with recombinant human CA1, 3, 5A, 5B, 6, 7, 8, and 10 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction