Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carbonic Anhydrase XII/CA12 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2190
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2190 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2190 Supplier R&D Systems Supplier No. AF2190
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carbonic Anhydrase XII/CA12 Polyclonal specifically detects Carbonic Anhydrase XII/CA12 in Human samples. It is validated for Western Blot, Simple Western.

Specifications

Antigen Carbonic Anhydrase XII/CA12
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 10 ug/mL
Gene Accession No. O43570
Gene Alias CA12, Carbonate dehydratase XII, carbonic anhydrase 12, carbonic anhydrase XIICAXII, carbonic dehydratase, CA-XII, EC 4.2.1.1, FLJ20151, HsT18816, Tumor antigen HOM-RCC-3.1.3
Gene Symbols CA12
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase XII/CA12 Ala25-Gln291 Accession # O43570
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 771
Test Specificity Detects human Carbonic Anhydrase XII/CA12 in direct ELISAs and Western blots. In Western blots, approximately 40% cross-reactivity with recombinant mouse Carbonic Anhydrase XII/CA12 is observed and less than 5% cross-reactivity with recombinant human CA1, 3, 5A, 5B, 6, 7, 8, and 10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.