Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxylesterase 1/CES1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4920SP
This item is not returnable.
View return policy
Description
Carboxylesterase 1/CES1 Monoclonal specifically detects Carboxylesterase 1/CES1 in Human samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.Specifications
Carboxylesterase 1/CES1 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 25 ug/mL | |
P23141 | |
CES1 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase1/CES-1 His19-Glu563 Accession # P23141 | |
25 μg | |
Primary | |
Detects human Carboxylesterase1/CES-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse (rm) CES-3 and no cross-reactivity with rmCES-2, rmCES-5, or recombinant human CES-2 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
544623 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACAT, acyl coenzyme A:cholesterol acyltransferase, Acyl-coenzyme A:cholesterol acyltransferase, Brain carboxylesterase hBr1, carboxylesterase 1, carboxylesterase 1 (monocyte/macrophage serine esterase 1), carboxylesterase 2 (liver), CEH, Ces-1, CES1, CES1A2, CES2EC 3.1.1.1, cholesteryl ester hydrolase, Cocaine carboxylesterase, EC 3.1.1, egasyn, HMSE1, HMSECES1A1, human monocyte/macrophage serine esterase 1, liver carboxylesterase 1, MGC117365, Monocyte/macrophage serine esterase, PCE-1, REH, Retinyl ester hydrolase, Serine esterase 1, Ses-1, SES1, SES1TGH, TGH, Triacylglycerol hydrolase | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1066 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction