Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxylesterase 2/CES2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $467.50
Specifications
| Antigen | Carboxylesterase 2/CES2 |
|---|---|
| Clone | 861429 |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB56571SP
![]() |
R&D Systems
MAB56571SP |
25 μg |
Each for $200.50
|
|
|||||
MAB56571100
![]() |
R&D Systems
MAB56571100 |
100 μg |
Each for $467.50
|
|
|||||
Description
Carboxylesterase 2/CES2 Monoclonal specifically detects Carboxylesterase 2/CES2 in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
| Carboxylesterase 2/CES2 | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| carboxylesterase 2 (intestine, liver), carboxylesterase 2EC 3.1.1.1, CE-2, CE-2PCE-2, CES2, CES2A1, EC 3.1.1, EC 3.1.1.84, hCE-2, iCE, intestinal carboxylesterase; liver carboxylesterase-2, PCE-2 | |
| Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase 2/CES2, Gln27-Leu559, Accession # O00748 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 861429 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| O00748 | |
| 8824 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title