Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxypeptidase A1/CPA1 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | Carboxypeptidase A1/CPA1 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.7 ug/mL |
Applications | Western Blot, Immunoprecipitation, Neutralization |
Classification | Polyclonal |
Description
Carboxypeptidase A1/CPA1 Polyclonal specifically detects Carboxypeptidase A1/CPA1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
Carboxypeptidase A1/CPA1 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 1.7 ug/mL | |
Polyclonal | |
Goat | |
Human | |
carboxypeptidase A1, carboxypeptidase A1 (pancreatic), CPA, CPA1, EC 3.4.17, EC 3.4.17.1 | |
CPA1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation, Neutralization | |
Unconjugated | |
RUO | |
P15085 | |
1357 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase A1/CPA1 Lys17-Tyr419 Accession # P15085 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human Carboxypeptidase A1/CPA1 in direct ELISAs and Western blots. In direct ELISAs, approximately 40% cross-reactivity with recombinant mouse CPA1 is observed and less than 10% cross-reactivity with recombinant human CPB1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title