Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxypeptidase A2/CPA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Carboxypeptidase A2/CPA2 |
---|---|
Clone | 384023 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB28961SP
![]() |
R&D Systems
MAB28961SP |
25 μg |
Each for $200.50
|
|
|||||
MAB28961
![]() |
R&D Systems
MAB28961 |
100 μg |
Each for $468.00
|
|
|||||
Description
Carboxypeptidase A2/CPA2 Monoclonal specifically detects Carboxypeptidase A2/CPA2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase A2/CPA2 | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P48052 | |
1358 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase A2/CPA2 Leu18-His417 Accession # P48052 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Carboxypeptidase A2/CPA2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CPA1, rhCPA4, rhCPB1, or rhCPE is observed. Recognizes the pro form (aa 17-417), but not the activated enzyme (aa 113-417) in Western blots. |
384023 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
carboxypeptidase A2, carboxypeptidase A2 (pancreatic), CPA2, EC 3.4.17, EC 3.4.17.15 | |
CPA2 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title