Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Carboxypeptidase B2/CPB2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6036
This item is not returnable.
View return policy
Description
Carboxypeptidase B2/CPB2 Monoclonal specifically detects Carboxypeptidase B2/CPB2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carboxypeptidase B2/CPB2 | |
Monoclonal | |
LYOPH | |
Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
Q96IY4 | |
CPB2 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxypeptidase B2/CPB2 Phe23-Val423 (predicted) Accession # Q96IY4 | |
100 μg | |
Primary | |
Detects human Carboxypeptidase B2/CPB2 in direct ELISAs and Western blots. In Western blots, approximately 20% cross-reactivity with recombinant human (rh) Carboxypeptidase A1, A4, recombinant mouse (rm) Carboxypeptidase A1, A4, B1, and 5% cross-reactivity with rhCarboxypeptidase B1, B2, and E is observed under reducing conditions. No cross-reactivity with any of these proteins is observed under nonreducing conditions. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
650801 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
carboxypeptidase B2, carboxypeptidase B2 (plasma), carboxypeptidase B2 (plasma, carboxypeptidase U), Carboxypeptidase U, CPB2, CPU, CPUEC 3.4.17.20, EC 3.4.17, PCPB, Plasma carboxypeptidase B, TAFI, TAFIcarboxypeptidase B-like protein, Thrombin-activable fibrinolysis inhibitor, thrombin-activatable fibrinolysis inhibitor | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1361 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction