Learn More
Human Carboxypeptidase X1/CPXM1 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Carboxypeptidase X1/CPXM1 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Gene Accession No. | Q96SM3 |
| Gene Alias | carboxypeptidase X (M14 family), carboxypeptidase X (M14 family), member 1, CPX1, CPX-1, CPX1EC 3.4.17.-, CPXM, CPXM1, EC 3.4.17, EC 3.4.17.10, EC 3.4.17.3, Metallocarboxypeptidase CPX-1, probable carboxypeptidase X1 |
| Gene Symbols | CPXM1 |
| Host Species | Goat |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.