Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cardiotrophin-1/CT-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB2602100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2602100 100 μg
MAB2602SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB2602100 Supplier R&D Systems Supplier No. MAB2602100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cardiotrophin-1/CT-1 Monoclonal specifically detects Cardiotrophin-1/CT-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.

Specifications

Antigen Cardiotrophin-1/CT-1
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone 89221
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, ELISA, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q16619
Gene Alias cardiophin 1, cardiotrophin 1, cardiotrophin-1, Cardiotrophin1, CT-1CT1, CTF1
Host Species Mouse
Immunogen E. coli-derived recombinant human Cardiotrophin-1 Ser2-Ala201 Accession # Q16619
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489
Test Specificity Detects human Cardiotrophin-1 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.