Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Carnosine Dipeptidase 1/CNDP1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB24891SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB24891SP 25 μg
MAB24891 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB24891SP Supplier R&D Systems Supplier No. MAB24891SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carnosine Dipeptidase 1/CNDP1 Monoclonal specifically detects Carnosine Dipeptidase 1/CNDP1 in Human samples. It is validated for Immunoprecipitation, Neutralization.

Specifications

Antigen Carnosine Dipeptidase 1/CNDP1
Applications Immunoprecipitation, Neutralization
Classification Monoclonal
Clone 352213
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunoprecipitation 25 ug/mL, Neutralization 0.6 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q96KN2
Gene Alias beta-Ala-His dipeptidase, carnosinase 1, Carnosine dipeptidase 1, carnosine dipeptidase 1 (metallopeptidase M20 family), CN1MGC102737, CNDP dipeptidase 1, CNDP1, CPGL2, CPGL2MGC142072, EC 3.4.13.20, Glutamate carboxypeptidase-like protein 2, HsT2308, MGC10825, Serum carnosinase
Gene Symbols CNDP1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Carnosine Dipeptidase 1/CNDP1 Pro28-His507 Accession # Q96KN2
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84735
Test Specificity Detects human Carnosine Dipeptidase 1/CNDP1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.