Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Caspase-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $432.50
Specifications
Antigen | Caspase-1 |
---|---|
Clone | 1021430 |
Dilution | Western Blot 2 ug/mL, Simple Western 20 ug/mL, ELISA, Knockout Validated |
Applications | Western Blot, ELISA, KnockDown |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB62151SP
![]() |
R&D Systems
MAB62151SP |
25 μg |
Each for $200.50
|
|
|||||
MAB62151100
![]() |
R&D Systems
MAB62151100 |
100 μg |
Each for $432.50
|
|
|||||
Description
Caspase-1 Monoclonal specifically detects Caspase-1 in Human samples. It is validated for Western Blot, Simple Western, ELISA, Knockout Validated.Specifications
Caspase-1 | |
Western Blot 2 ug/mL, Simple Western 20 ug/mL, ELISA, Knockout Validated | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS. | |
CASP1, CASP-1, CASP1 nirs variant 1, caspase 1, apoptosis-related cysteine peptidase, caspase-1, Caspase1, Caspase-1 subunit p10, Caspase-1 subunit p20, EC 3.4.22.36, EC:3.4.22.36, ICE, IL-1 beta-converting enzyme, IL1BC, IL-1BC, IL1BCE, IL1B-convertase, interleukin 1, beta, convertase, interleukin 1-B converting enzyme, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45 | |
E. coli-derived recombinant human Caspase-1, Asn120-Asp297, Accession # P29466 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
1021430 | |
Western Blot, ELISA, KnockDown | |
Unconjugated | |
Mouse | |
Human | |
P29466 | |
834 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title