Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                Human Caspase-1 Antibody, R&D Systems™
                                
                                
                                
                                
                            
                            
                            
                                
                                    
Rabbit Monoclonal Antibody
$200.50 - $472.87
Specifications
| Antigen | Caspase-1 | 
|---|---|
| Clone | 2666E | 
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL | 
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry | 
| Classification | Monoclonal | 
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
                
                
                    
                        MAB62156SP
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        R&D Systems
                         MAB62156SP  | 
                
            
            
            
            
            
                
                    25 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $200.50
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
                
                
                    
                        MAB62156100
                    
                    
                        
                    
                    
                        ![]()  | 
            
                
                    
                        R&D Systems
                         MAB62156100  | 
                
            
            
            
            
            
                
                    100 μg | 
                    
                        
                        
                            
                             
                                        
                                            
                                                
                                                
                                                
                                            
                                        
                                        
                                            
                                            
                                                
                                                
                                            
                                        
                                        
                                            
                                                
                                                Each for $472.87
                                                 
                                
                             | 
                
                    
                        
                             | 
                    
                
                
                    |||||
Description
Caspase-1 Monoclonal antibody specifically detects Caspase-1 in Human samples. It is validated for Western Blot, Immunohistochemistry, ImmunocytochemistrySpecifications
| Caspase-1 | |
| Western Blot 2 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CASP1, CASP-1, CASP1 nirs variant 1, caspase 1, apoptosis-related cysteine peptidase, caspase-1, Caspase1, Caspase-1 subunit p10, Caspase-1 subunit p20, EC 3.4.22.36, EC:3.4.22.36, ICE, IL-1 beta-converting enzyme, IL1BC, IL-1BC, IL1BCE, IL1B-convertase, interleukin 1, beta, convertase, interleukin 1-B converting enzyme, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45 | |
| E. coli-derived recombinant human Caspase-1, Asn120-Asp297, Accession # P29466 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | 
| 2666E | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P29466 | |
| 834 | |
| IgG | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | 
Spot an opportunity for improvement?Share a Content Correction
            
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title