Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Caspase-1 ELISA Kit (Chemiluminescence)
SDP

Catalog No. NB294678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294678 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294678 Supplier Novus Biologicals™ Supplier No. NBP325941

Specifications

Assay Range 6.25 to 400 pg/mL
Assay Sensitivity 3.75 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Other Biological Fluids
Specificity This kit recognizes Human Caspase-1 in samples. No significant cross-reactivity or interference between Human Caspase-1 and analogues was observed.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 834
Quantity 1 Kit
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym CASP1, CASP-1, CASP1 nirs variant 1, caspase 1, apoptosis-related cysteine peptidase, caspase-1, Caspase-1 subunit p10, Caspase-1 subunit p20, EC 3.4.22.36, EC:3.4.22.36, ICE, IL-1 beta-converting enzyme, IL1BC, IL-1BC, IL1BCE, IL1B-convertase, interleukin 1, beta, convertase, interleukin 1-B converting enzyme, Interleukin-1 beta convertase, Interleukin-1 beta-converting enzyme, p45
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.