Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Caspase-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB7228
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB7228 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB7228 Supplier R&D Systems Supplier No. MAB7228
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Caspase-2 Monoclonal specifically detects Caspase-2 in Human samples. It is validated for Immunocytochemistry.

Specifications

Antigen Caspase-2
Applications Immunocytochemistry
Classification Monoclonal
Clone 691233
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunocytochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P42575
Gene Alias caspase-2, CASP2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, Caspase2, ICH1, ICH-1, NEDD2, NEDD-2, PPP1R57
Gene Symbols CASP2
Host Species Mouse
Immunogen E. coli-derived recombinant human Caspase-2 Gly170-Asp333 & Ala348-Tyr452 Accession # P42575
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 835
Test Specificity Detects human Caspase-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Caspase-8 is observed.
Reconstitution Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.