Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Caspase-9 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8301
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8301 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB8301 Supplier R&D Systems Supplier No. MAB8301
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 6 publications

Caspase-9 Monoclonal specifically detects Caspase-9 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Caspase-9
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone LAP6
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Alias APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP9, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, Caspase9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease
Gene Symbols CASP9
Host Species Mouse
Immunogen E. coli-derived recombinant human Caspase-9 aa 1-134
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 842
Test Specificity Detects human Caspase-9 in Western blots and captures Caspase-9 complexed with APAF-1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.