Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Caspase-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: R&D Systems MAB8301SP
This item is not returnable.
View return policy
Description
Caspase-9 Monoclonal specifically detects Caspase-9 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.Specifications
Caspase-9 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL | |
APAF-3APAF3, apoptotic protease activating factor 3, Apoptotic protease Mch-6, Apoptotic protease-activating factor 3, CASP9, CASP-9, caspase 9, apoptosis-related cysteine peptidase, caspase-9, Caspase9, EC 3.4.22.62, ICE-LAP6CASPASE-9c, ICE-like apoptotic protease 6, MCH6apoptosis-related cysteine protease | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
842 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot, Immunohistochemistry | |
LAP6 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CASP9 | |
E. coli-derived recombinant human Caspase-9 aa 1-134 | |
25 μg | |
Primary | |
Detects human Caspase-9 in Western blots and captures Caspase-9 complexed with APAF-1. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction