Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Caspr2 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$200.50 - $432.50

Specifications

Antigen Caspr2
Clone 840727
Dilution Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Applications Flow Cytometry, CyTOF
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB5145SP
GREENER_CHOICE
View Documents
R&D Systems
MAB5145SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB5145100
GREENER_CHOICE
View Documents
R&D Systems
MAB5145100
100 μg
Each for $432.50
Only null left
Add to Cart
 
Description

Description

Caspr2 Monoclonal specifically detects Caspr2 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications

Specifications

Caspr2
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Monoclonal
Purified
RUO
Lyophilized from a 0.2 µm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 µm filtered solution in PBS.
AUTS15, CASPR2DKFZp781D1846, CDFE, Cell recognition molecule Caspr2, CNTNAP2, contactin associated protein-like 2, contactin-associated protein 2, contactin-associated protein-like 2, homolog of Drosophila neurexin IV, KIAA0868, NRXN4, NRXN4Caspr2, PTHSL1
Mouse myeloma cell line NS0-derived recombinant human Caspr2, Pro29-Ala1262, Accession # Q9UHC6
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human Caspr2 in direct ELISAs.
840727
Flow Cytometry, CyTOF
Unconjugated
Mouse
Human
Q9UHC6
26047
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.