Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Catalase Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3398SP
This item is not returnable.
View return policy
Description
Catalase Monoclonal specifically detects Catalase in Human samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.Specifications
Catalase | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 μg/mL, Simple Western 5 μg/mL, Immunocytochemistry 3-25 μg/mL, Knockout Validated | |
P04040 | |
CAT | |
E. coli-derived recombinant human Catalase Met1-Leu527 Accession # P04040 | |
25 μg | |
Primary | |
Detects human Catalase in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse Catalase or recombinant human Serpin C1 is observed. | |
Human | |
Purified |
Western Blot, Immunocytochemistry, Immunoassay | |
724810 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Cas1, CAT, catalase, Cs-1, EC 1.11.1.6, MGC138422, MGC138424 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
847 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction