Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Catalase ELISA Kit (Colorimetric)
SDP

Catalog No. NB170549 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB170549 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB170549 Supplier Novus Biologicals™ Supplier No. NBP318703
Add to Cart
Add to Cart
This item is not returnable. View return policy

Colorimetric ELISA Kit

Specifications

Assay Range 0.156 - 10 ng/mL (example only; lot dependent)
Assay Sensitivity 0.13 ng/mL (example only; lot dependent)
Product Type ELISA Kit (Colorimetric)
Sample Type Breast Milk, Cell Culture, Cerebrospinal Fluid, Plasma, Saliva, Serum, Tissue Extracts
For Use With (Application) ELISA
Gene ID (Entrez) 847
Gene Symbol CAT
Quantity 1 Kit
Research Discipline Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction
Storage Requirements Storage is content dependent.
Synonym catalase, EC 1.11.1.6, MGC138422, MGC138424
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.