Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cathepsin A/Lysosomal Carboxypeptidase A Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $667.00
Specifications
| Antigen | Cathepsin A/Lysosomal Carboxypeptidase A |
|---|---|
| Clone | 179803 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
Description
Cathepsin A/Lysosomal Carboxypeptidase A Monoclonal specifically detects Cathepsin A/Lysosomal Carboxypeptidase A in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Cathepsin A/Lysosomal Carboxypeptidase A | |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Human | |
| P10619 | |
| 5476 | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin A/Lysosomal Carboxypeptidase A Ala29-Tyr480 (predicted) Accession # P10619 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Cathepsin A/Lysosomal Carboxypeptidase A in direct ELISAs and Western blots. In Western blots, detects the single chain (55 kDa) and heavy chain (32 kDa) forms of recombinant human (rh) Cathepsin A. In Western blots, less than 5% cross-reactivity with rhCathepsin B, C, D, E, L, O, S, X and Z is observed and no cross-reactivity with the light chain (20 kDa) of rhCathepsin A is observed. |
| 179803 | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| beta-galactosidase 2, beta-galactosidase protective protein, Carboxypeptidase C, Carboxypeptidase L, cathepsin ANGBE, CTSA, EC 3.4.16, EC 3.4.16.5, GLB2, GSL, Lysosomal Carboxypeptidase A, lysosomal protective protein, PPCA, PPGBprotective protein for beta-galactosidase (galactosialidosis), Protective protein cathepsin A, Protective protein for beta-galactosidase | |
| CTSA | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title