Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cathepsin D Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1014
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1014 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB1014 Supplier R&D Systems Supplier No. MAB1014
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cathepsin D Monoclonal specifically detects Cathepsin D in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen Cathepsin D
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 185111
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Simple Western 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P07339
Gene Alias cathepsin D, cathepsin D (lysosomal aspartyl protease), CPSD, CTSD, EC 3.4.23, EC 3.4.23.5, lysosomal aspartyl peptidase, lysosomal aspartyl protease, MGC2311, neuronal 10
Gene Symbols CTSD
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cathepsin D Leu21-Leu412 Accession # P07339
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1509
Test Specificity Detects human Cathepsin D in direct ELISAs and Western blots. It recognizes both the pro and mature forms of recombinant human (rh) Cathepsin D. In direct ELISAs, no cross-reactivity with rhCathepsin A, rhCathepsin B, rhCathepsin C, rhCathepsin L, rhCathepsin O, rhCathepsin S, rhCathepsin Z, or recombinant mouse Cathepsin D is observed. In Western blots, 100% cross-reactivity with rhCathepsin E and rmCathepsin D is observed and no cross-reactivity with rhBACE-1 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.