Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cathepsin E Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
$200.50 - $526.50
Specifications
Antigen | Cathepsin E |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
Classification | Polyclonal |
Description
Cathepsin E Polyclonal specifically detects Cathepsin E in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
Cathepsin E | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Human | |
CATE, cathepsin E, CTSE, EC 3.4.23, EC 3.4.23.34, erythrocyte membrane aspartic proteinase, slow-moving proteinase | |
CTSE | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Unconjugated | |
RUO | |
NP_001901 | |
1510 | |
Mouse myeloma cell line NS0-derived recombinant human Cathepsin E Ile54-Pro396 Accession # NP_001901 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human pro and active Cathepsin E in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 40% cross-reactivity with recombinant mouse Cathepsin E is observed and less than 2% cross-reactivity with recombinant human (rh) Cathepsin D and rhBACE is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title