Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cathepsin F Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2075SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2075SP 25 μg
AF2075 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2075SP Supplier R&D Systems Supplier No. AF2075SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Cathepsin F Polyclonal specifically detects Cathepsin F in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen Cathepsin F
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9UBX1
Gene Alias cathepsin F, CATSFEC 3.4.22.41, CTSF, EC 3.4.22
Gene Symbols CTSF
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Cathepsin F Ala20-Asp484 Accession # Q9UBX1
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8722
Test Specificity Detects human Cathepsin F in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse Cathepsin F is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.