Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cathepsin H Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7516SP
This item is not returnable.
View return policy
Description
Cathepsin H Polyclonal specifically detects Cathepsin H in Human samples. It is validated for Western Blot.Specifications
Cathepsin H | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACC-4, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin H, CPSB, CTSH, DKFZp686B24257, EC 3.4.22, EC 3.4.22.16, MGC1519, minichain, N-benzoylarginine-beta-naphthylamide hydrolase | |
Sheep | |
Affinity Purified | |
RUO | |
1512 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoassay | |
LYOPH | |
Western Blot 0.2 ug/mL, Knockout Validated | |
CAA34734 | |
CTSH | |
Chinese hamster ovary cell line CHO-derived recombinant human Cathepsin H Ala23-Val335 Accession # CAA34734 | |
25 μg | |
Primary | |
Detects human Cathepsin H in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse Cathepsin H is observed, and less than 1% cross-reactivity with recombinant human Cathepsin L2 is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction