Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cathepsin H Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7516
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7516 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7516 Supplier R&D Systems Supplier No. AF7516
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Cathepsin H Polyclonal specifically detects Cathepsin H in Human samples. It is validated for Western Blot.

Specifications

Antigen Cathepsin H
Applications Western Blot, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.2 ug/mL, Knockout Validated
Gene Accession No. CAA34734
Gene Alias ACC-4, ACC-5, aleurain, cathepsin B3, cathepsin BA, cathepsin H, CPSB, CTSH, DKFZp686B24257, EC 3.4.22, EC 3.4.22.16, MGC1519, minichain, N-benzoylarginine-beta-naphthylamide hydrolase
Gene Symbols CTSH
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human Cathepsin H Ala23-Val335 Accession # CAA34734
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1512
Test Specificity Detects human Cathepsin H in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant mouse Cathepsin H is observed, and less than 1% cross-reactivity with recombinant human Cathepsin L2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.