Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cathepsin O Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB1174
This item is not returnable.
View return policy
Description
Cathepsin O Monoclonal specifically detects Cathepsin O in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| Cathepsin O | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| P43234 | |
| CTSO | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin O Asp24-Val321 Accession # P43234 | |
| 500 μg | |
| Primary | |
| Detects human Cathepsin O in Western blots and direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human Cathepsin A, B, C, D, E, L, L2, S, Z, or recombinant mouse Cathepsin H is observed. | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 180406 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cathepsin O, CTSO, CTSO1, EC 3.4.22.42 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1519 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction