Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Cbx2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8098SP
This item is not returnable.
View return policy
Description
Cbx2 Monoclonal specifically detects Cbx2 in Human samples. It is validated for Immunocytochemistry.Specifications
Cbx2 | |
Monoclonal | |
LYOPH | |
Immunocytochemistry 8-25 ug/mL | |
Q14781 | |
CBX2 | |
E. coli-derived recombinant human Cbx2 Lys131-His217 Accession # Q14781 | |
25 μg | |
Primary | |
Detects human Cbx2 in ELISA. | |
Human | |
Purified |
Immunocytochemistry | |
880833 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CDCA6, cell division cycle associated 6, chromobox homolog 2, chromobox homolog 2 (Drosophila Pc class), chromobox homolog 2 (Pc class homolog, Drosophila), chromobox protein homolog 2, M33, MGC10561, modifier 3, Pc class homolog, SRXY5 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
84733 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction