Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CC Chemokine Receptor D6 Alexa Fluor™ 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1364R100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1364R100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1364R100 Supplier R&D Systems Supplier No. FAB1364R100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

CC Chemokine Receptor D6 Monoclonal specifically detects CC Chemokine Receptor D6 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen CC Chemokine Receptor D6
Applications Flow Cytometry
Classification Monoclonal
Clone 196124
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. O00590
Gene Alias ACKR2, C-C chemokine receptor D6, CCBP2, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250
Gene Symbols CCBP2
Host Species Rat
Immunogen RBL-1 rat basophilic leukemia cell line transfected with human CC Chemokine Receptor D6 Met1-Ala384 Accession # O00590
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1238
Test Specificity Detects human CC Chemokine Receptor D6 in transfectants. Does not stain the parent cell line.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.