Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CC Chemokine Receptor D6 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
Supplier: R&D Systems MAB1364SP
This item is not returnable.
View return policy
Description
CC Chemokine Receptor D6 Monoclonal specifically detects CC Chemokine Receptor D6 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
CC Chemokine Receptor D6 | |
Monoclonal | |
LYOPH | |
Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
O00590 | |
CCBP2 | |
RBL-1 rat basophilic leukemia cell line transfected with human CC Chemokine Receptor D6 Met1-Ala384 Accession # O00590 | |
25 μg | |
Primary | |
Detects human CC Chemokine Receptor D6 in transfectants. Does not stain the parent cell line. | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
196124 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
ACKR2, C-C chemokine receptor D6, CCBP2, CCR10CCR9D6CC-chemokine-binding receptor JAB61, chemokine (C-C) receptor 9, chemokine binding protein 2, Chemokine receptor CCR-10, Chemokine receptor CCR-9, chemokine receptor D6, chemokine-binding protein 2, Chemokine-binding protein D6, CMKBR9, CMKBR9chemokine (C-C motif) receptor 9, hD6, MGC126678, MGC138250 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1238 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction